ENAH303681E3 Enamine

4-methanesulfonyl-3-methylphenol

  • MDL number MFCD00464310
  • Cas Number : 14270-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729907
Availability details for
Quantity
  • Availability
  • Add to Cart