ENAH303681E5 Enamine

2-{[(tert-butoxy)carbonyl]amino}-3-(methylsulfanyl)butanoic acid

  • MDL number MFCD31719575
  • Cas Number : 2253639-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729925
Availability details for
Quantity
  • Availability
  • Add to Cart