ENAH303681E8 Enamine

ethyl 2-[(1E)-2,2-difluorocycloheptylidene]acetate

  • Cas Number : 2253644-38-9
  • Molecular Weight : 218.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (E)-2-(2,2-difluorocycloheptylidene)acetate
Inchi Code 1S/C11H16F2O2/c1-2-15-10(14)8-9-6-4-3-5-7-11(9,12)13/h8H,2-7H2,1H3/b9-8+
Inchi Key HAEIMQHUGKZEKD-CMDGGOBGSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6729936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729936
Availability details for
Quantity
  • Availability
  • Add to Cart