ENAH303681E9 Enamine

2-[(1E)-2,2-difluorocycloheptylidene]acetic acid

  • MDL number MFCD31719618
  • Cas Number : 2253644-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729937
Availability details for
Quantity
  • Availability
  • Add to Cart