ENAH303681EA Enamine

3-(difluoromethyl)-4-methoxyaniline hydrochloride

  • MDL number MFCD31719577
  • Cas Number : 2253630-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729940
Availability details for
Quantity
  • Availability
  • Add to Cart