ENAH303681F9 Enamine

benzyl 1,1-dioxo-hexahydro-1lambda6-[1,2,3]oxathiazolo[3,4-a]piperazine-5-carboxylate

  • MDL number MFCD31719624
  • Cas Number : 2253639-04-0
  • Molecular Weight : 312.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl tetrahydro-[1,2,3]oxathiazolo[3,4-a]pyrazine-5(3H)-carboxylate 1,1-dioxide
Inchi Code 1S/C13H16N2O5S/c16-13(19-9-11-4-2-1-3-5-11)14-6-7-15-12(8-14)10-20-21(15,17)18/h1-5,12H,6-10H2
Inchi Key OTPJBUGDTDJFRF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730144
Availability details for
Quantity
  • Availability
  • Add to Cart