ENAH303681FD Enamine

1-(5-tert-butyl-1,3,4-thiadiazol-2-yl)-3-methylurea

  • MDL number MFCD00177853
  • Cas Number : 24814-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-210
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730156
Availability details for
Quantity
  • Availability
  • Add to Cart