ENAH30368201 Enamine

4-(3-chlorobenzenesulfinyl)butanenitrile

  • MDL number MFCD20394687
  • Cas Number : 1342845-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730162
Availability details for
Quantity
  • Availability
  • Add to Cart