ENAH30368202 Enamine

(E)-2-(2,4-dichlorophenyl)ethene-1-sulfonyl chloride

  • Cas Number : 62755-57-1
  • Molecular Weight : 271.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (E)-2-(2,4-dichlorophenyl)ethenesulfonyl chloride
Inchi Code 1S/C8H5Cl3O2S/c9-7-2-1-6(8(10)5-7)3-4-14(11,12)13/h1-5H/b4-3+
Inchi Key MQFAGVDNFKYFHR-ONEGZZNKSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730171

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6730171
pictograms GHS05,GHS07
hstatements H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart