ENAH30368205 Enamine

2-(piperidin-4-yl)-1,3-oxazole-4-carboxylic acid hydrochloride

  • MDL number MFCD31719594
  • Cas Number : 2253632-66-3
  • Molecular Weight : 232.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(piperidin-4-yl)oxazole-4-carboxylic acid hydrochloride
Inchi Code 1S/C9H12N2O3.ClH/c12-9(13)7-5-14-8(11-7)6-1-3-10-4-2-6;/h5-6,10H,1-4H2,(H,12,13);1H
Inchi Key AKODCPLGSODXKC-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730184

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6730184
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart