ENAH30368206 Enamine

N-(1H-pyrazol-4-yl)pyridazin-3-amine hydrochloride

  • MDL number MFCD31719595
  • Cas Number : 2253630-45-2
  • Molecular Weight : 197.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(1H-pyrazol-4-yl)pyridazin-3-amine hydrochloride
Inchi Code 1S/C7H7N5.ClH/c1-2-7(12-8-3-1)11-6-4-9-10-5-6;/h1-5H,(H,9,10)(H,11,12);1H
Inchi Key DUVUUXRWYCCJDL-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730205
Availability details for
Quantity
  • Availability
  • Add to Cart