ENAH30368207 Enamine

2-amino-3-[5-(trifluoromethyl)thiophen-3-yl]propanoic acid hydrochloride

  • MDL number MFCD31719421
  • Cas Number : 2253638-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730212
Availability details for
Quantity
  • Availability
  • Add to Cart