ENAH30368208 Enamine

(2E)-5,5,6,6,7,7,7-heptafluoro-2-methoxyhept-2-en-4-one

  • Cas Number : 1246896-27-4
  • Molecular Weight : 268.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (E)-5,5,6,6,7,7,7-heptafluoro-2-methoxyhept-2-en-4-one
Inchi Code 1S/C8H7F7O2/c1-4(17-2)3-5(16)6(9,10)7(11,12)8(13,14)15/h3H,1-2H3/b4-3+
Inchi Key AZDYPLIEDNDVMP-ONEGZZNKSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730220
Availability details for
Quantity
  • Availability
  • Add to Cart