ENAH30368209 Enamine

(1E)-1-ethoxy-4,4,5,5,6,6,6-heptafluoro-2-methylhex-1-en-3-one

  • MDL number MFCD31719422
  • Cas Number : 1414887-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730221
Availability details for
Quantity
  • Availability
  • Add to Cart