ENAH3036820A Enamine

(4E)-1,1,1,2,2-pentafluoro-5-methoxyhex-4-en-3-one

  • MDL number MFCD07368412
  • Cas Number : 934299-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730223
Availability details for
Quantity
  • Availability
  • Add to Cart