ENAH3036820C Enamine

ethyl 6-bromo-1,2,3,4-tetrahydroisoquinoline-1-carboxylate hydrochloride

  • MDL number MFCD11858423
  • Cas Number : 1260637-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730226
Availability details for
Quantity
  • Availability
  • Add to Cart