ENAH3036820D Enamine

(2S,4S)-4-phenoxypyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD31719424

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730227
Availability details for
Quantity
  • Availability
  • Add to Cart