ENAH30368213 Enamine

N-(3-aminopropyl)cyclopropanesulfonamide hydrochloride

  • MDL number MFCD31719427
  • Cas Number : 2253638-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730275
Availability details for
Quantity
  • Availability
  • Add to Cart