ENAH30368215 Enamine

3-(azetidin-3-ylidene)pyrrolidine-2,5-dione hydrochloride

  • MDL number MFCD31719657
  • Cas Number : 2253639-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730277
Availability details for
Quantity
  • Availability
  • Add to Cart