ENAH30368218 Enamine

3',4'-dihydro-1'H-spiro[azetidine-3,2'-quinazolin]-4'-one hydrochloride

  • MDL number MFCD31719659
  • Cas Number : 2253632-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730281
Availability details for
Quantity
  • Availability
  • Add to Cart