ENAH30368219 Enamine

1-(5-bromo-1H-indol-3-yl)-2-methylpropan-2-amine hydrochloride

  • MDL number MFCD31719660
  • Cas Number : 2253632-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730287
Availability details for
Quantity
  • Availability
  • Add to Cart