ENAH3036821D Enamine

methyl 3-iodo-4-(propan-2-yl)benzoate

  • MDL number MFCD11112911
  • Molecular Weight : 304.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-iodo-4-isopropylbenzoate
Inchi Code 1S/C11H13IO2/c1-7(2)9-5-4-8(6-10(9)12)11(13)14-3/h4-7H,1-3H3
Inchi Key VIUQYXQVBWXXBV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730302
Availability details for
Quantity
  • Availability
  • Add to Cart