ENAH30368220 Enamine

2-(difluoromethoxy)quinolin-7-ol

  • MDL number MFCD18416066
  • Cas Number : 1261475-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730311
Availability details for
Quantity
  • Availability
  • Add to Cart