ENAH30368221 Enamine

1-(4-fluorophenyl)-1-(5-methyl-1,2,4-oxadiazol-3-yl)methanamine hydrochloride

  • MDL number MFCD31719662
  • Cas Number : 2253641-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730324
Availability details for
Quantity
  • Availability
  • Add to Cart