ENAH30368223 Enamine

2,7-bis(difluoromethoxy)quinoline

  • MDL number MFCD31719598
  • Cas Number : 2253641-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730347
Availability details for
Quantity
  • Availability
  • Add to Cart