ENAH30368226 Enamine

6,8-dichloro-2-ethylimidazo[1,2-b]pyridazine

  • MDL number MFCD31719664
  • Cas Number : 2253640-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730354
Availability details for
Quantity
  • Availability
  • Add to Cart