ENAH30368228 Enamine

3-(thian-4-yl)azetidine hydrochloride

  • MDL number MFCD31719600
  • Cas Number : 2253632-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730357
Availability details for
Quantity
  • Availability
  • Add to Cart