ENAH3036822A Enamine

tert-butyl 4-(2-aminophenyl)-3-oxopiperazine-1-carboxylate

  • MDL number MFCD30490555
  • Cas Number : 2253630-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730360
Availability details for
Quantity
  • Availability
  • Add to Cart