ENAH3036822C Enamine

2-[4-(butan-2-yloxy)-3,5-dimethoxyphenyl]ethan-1-amine

  • MDL number MFCD12870975
  • Cas Number : 1183789-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730383
Availability details for
Quantity
  • Availability
  • Add to Cart