ENAH3036822E Enamine

4-(1H-1,2,3-triazol-1-yl)benzaldehyde

  • MDL number MFCD07783843
  • Cas Number : 41498-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730388
Availability details for
Quantity
  • Availability
  • Add to Cart