ENAH3036822F Enamine

2-(3,5-dibromopyridin-4-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD31719665
  • Cas Number : 2253638-49-0
  • Molecular Weight : 352.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,5-dibromopyridin-4-yl)ethan-1-amine dihydrochloride
Inchi Code 1S/C7H8Br2N2.2ClH/c8-6-3-11-4-7(9)5(6)1-2-10;;/h3-4H,1-2,10H2;2*1H
Inchi Key ROVBYAGFXSIJOA-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730404
Availability details for
Quantity
  • Availability
  • Add to Cart