ENAH30368230 Enamine

4-chloro-6-methanesulfonylpyrimidine

  • MDL number MFCD14702659
  • Cas Number : 89283-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-127
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730411
Availability details for
Quantity
  • Availability
  • Add to Cart