ENAH30368232 Enamine

5-methyl-2,3,4,5-tetrahydro-1H-3-benzazepin-7-ol hydrobromide

  • MDL number MFCD31719666
  • Cas Number : 2253629-68-2
  • Molecular Weight : 258.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-2,3,4,5-tetrahydro-1H-benzo[d]azepin-7-ol hydrobromide
Inchi Code 1S/C11H15NO.BrH/c1-8-7-12-5-4-9-2-3-10(13)6-11(8)9;/h2-3,6,8,12-13H,4-5,7H2,1H3;1H
Inchi Key CIZVQFQBXGEKCV-UHFFFAOYSA-N

Description

Purity 95
Salt Data Br
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730438
Availability details for
Quantity
  • Availability
  • Add to Cart