ENAH30368236 Enamine

spiro[5.5]undecan-2-amine hydrochloride

  • MDL number MFCD31719668
  • Cas Number : 62439-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730451
Availability details for
Quantity
  • Availability
  • Add to Cart