ENAH3036823B Enamine

2-chloro-4-isocyanatoquinoline

  • MDL number MFCD18251098
  • Cas Number : 1260671-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730457
Availability details for
Quantity
  • Availability
  • Add to Cart