ENAH30368244 Enamine

2-[5-(phenoxymethyl)-1,2,4-oxadiazol-3-yl]ethan-1-amine hydrochloride

  • MDL number MFCD31719672
  • Cas Number : 2253632-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730520
Availability details for
Quantity
  • Availability
  • Add to Cart