ENAH3036824B Enamine

1-{5-[3-(hydroxymethyl)phenyl]thiophen-2-yl}ethan-1-ol

  • MDL number MFCD31719675
  • Cas Number : 2253640-70-7
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-(3-(hydroxymethyl)phenyl)thiophen-2-yl)ethan-1-ol
Inchi Code 1S/C13H14O2S/c1-9(15)12-5-6-13(16-12)11-4-2-3-10(7-11)8-14/h2-7,9,14-15H,8H2,1H3
Inchi Key SXPQSYSNDLJGSE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730620

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6730620
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart