ENAH30368253 Enamine

4,4-dimethyl-1-oxa-3,7-diazaspiro[4.4]nonan-2-one hydrochloride

  • MDL number MFCD31719406
  • Cas Number : 2253638-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730640
Availability details for
Quantity
  • Availability
  • Add to Cart