ENAH30368254 Enamine

4-chloro-2,3-difluorobenzene-1-sulfonyl chloride

  • MDL number MFCD28750492
  • Cas Number : 1805224-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730642
Availability details for
Quantity
  • Availability
  • Add to Cart