ENAH30368258 Enamine

4-(difluoromethylidene)cyclohexan-1-amine hydrochloride

  • MDL number MFCD31719607
  • Cas Number : 2253632-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730678
Availability details for
Quantity
  • Availability
  • Add to Cart