ENAH30368259 Enamine

(2,3-dihydro-1H-inden-1-ylidene)hydrazine

  • Cas Number : 5736-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-78
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730683
Availability details for
Quantity
  • Availability
  • Add to Cart