ENAH3036825A Enamine

2-chloro-4-cyclopentyl-6-(propan-2-yl)pyrimidine

  • MDL number MFCD22836651
  • Cas Number : 1412953-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730686

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730686
Availability details for
Quantity
  • Availability
  • Add to Cart