ENAH3036825B Enamine

ethyl 4-amino-2-fluorobutanoate hydrochloride

  • MDL number MFCD31719608
  • Cas Number : 2253630-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730692
Availability details for
Quantity
  • Availability
  • Add to Cart