ENAH3036825D Enamine

7-bromoquinolin-6-ol hydrobromide

  • MDL number MFCD31719409
  • Cas Number : 2253632-33-4
  • Molecular Weight : 304.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromoquinolin-6-ol hydrobromide
Inchi Code 1S/C9H6BrNO.BrH/c10-7-5-8-6(4-9(7)12)2-1-3-11-8;/h1-5,12H;1H
Inchi Key BDYZJNQJTBYRRI-UHFFFAOYSA-N

Description

Purity 95
Salt Data Br
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730696

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6730696
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart