ENAH3036825F Enamine

1-(pyrrolidin-3-yl)azetidin-3-ol; bis(trifluoroacetic acid)

  • MDL number MFCD30726877
  • Cas Number : 2206969-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2CF3COOH
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730698
Availability details for
Quantity
  • Availability
  • Add to Cart