ENAH30368260 Enamine

1-(5-fluoro-3-phenyl-1-benzofuran-2-yl)methanamine hydrochloride

  • MDL number MFCD31719610
  • Cas Number : 2253630-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730701
Availability details for
Quantity
  • Availability
  • Add to Cart