ENAH30368262 Enamine

1-methyl-2-(trifluoromethyl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD31719611
  • Cas Number : 2253640-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730703
Availability details for
Quantity
  • Availability
  • Add to Cart