ENAH30368264 Enamine

1-[(tert-butoxy)carbonyl]-3-(trifluoromethyl)piperidine-3-carboxylic acid

  • MDL number MFCD31719612
  • Cas Number : 2253638-97-8
  • Molecular Weight : 297.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tert-butoxycarbonyl)-3-(trifluoromethyl)piperidine-3-carboxylic acid
Inchi Code 1S/C12H18F3NO4/c1-10(2,3)20-9(19)16-6-4-5-11(7-16,8(17)18)12(13,14)15/h4-7H2,1-3H3,(H,17,18)
Inchi Key FIJSWJREJOOPEA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730768

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6730768
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart