ENAH30368267 Enamine

[(6-chloropyridin-3-yl)methyl](methyl)(propan-2-yl)amine

  • MDL number MFCD09809680
  • Cas Number : 1016688-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730787
Availability details for
Quantity
  • Availability
  • Add to Cart