ENAH30368268 Enamine

1-(chloromethyl)-3-(propylsulfanyl)benzene

  • MDL number MFCD31719410
  • Cas Number : 99047-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730797
Availability details for
Quantity
  • Availability
  • Add to Cart